Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03655424_s1
          See other LOC284630 GE Assays ›
          Gene Symbol:
          LOC284630
          Gene Name:
          uncharacterized LOC284630
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03655424_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC284630
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK096384.1 - 1 3151 113
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB325045.1 - 1 Chr. 1 - 2636352 - 2636897 [+] on Build GRCh38 113
          GenBankmRNA BM974150.1 - 1 Chr. 1 - 2636334 - 2637103 [+] on Build GRCh38 113
          GenBankmRNA CB243156.1 - 1 Chr. 1 - 2635976 - 2636670 [+] on Build GRCh38 113

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tetratricopeptide repeat domain 34 TTC34 Sense Chr. 1 - 2636988 - 2789737 on NCBI Build GRCh38
          Downstream actin related protein T2 ACTRT2 ARPM2,
          ARPT2,
          Arp-T2,
          HARPM2
          381.8kb Anti-sense Chr. 1 - 3021482 - 3022903 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013092 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20013115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00006025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15012615 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 111482 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 951339 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 494195 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 130335 AB014539.3 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 130335 AL583912.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 130335 NM_015254.3 refseq_mrna_human_NM
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3655775 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 22500 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3655775 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00002022 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00002022 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00002022 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1565728_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100006589.hg.1 ENST00000427302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006589.hg.1 ENST00000427302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006589.hg.1 ENST00000427302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006589.hg.1 ENST00000427302.1 Ensembl
          Human Genome U133 Plus 2.0 Array 239881_PM_at - -
          Human Genome U133 Plus 2.0 Array 1565729_PM_at - -
          Human Genome U133 Plus 2.0 Array 1565728_PM_at - -
          Human Genome U133 Plus 2.0 Array 239881_at - -
          Human Genome U133 Plus 2.0 Array 1565729_at - -
          Human Genome U133 Plus 2.0 Array 1565728_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565728_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1565729_at6 - -
          Human Genome U95 Set 70514_at - -
          Human Genome U95 Set 70514_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0