Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03655647_m1
          See other LOC729173 GE Assays ›
          Gene Symbol:
          LOC729173
          Gene Name:
          uncharacterized LOC729173
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 120168544 - 120171679 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03655647_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC729173
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK096981.1 - 1 - 2 178 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA921453.1 - 1 - 2 179 Chr. 11 - 120168977 - 120170153 [+] on Build GRCh38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC102724301 LOC102724301 -429.4kb Sense Chr. 11 - 119729583 - 119739623 on NCBI Build GRCh38
          Downstream out at first homolog OAF NS5ATP13TP2 39.4kb Sense Chr. 11 - 120211039 - 120229941 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK096981.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16509309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16509313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16509308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28499015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28499008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28499013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498951 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28499016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28499005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16509299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08071565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28498921 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P264063 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 26211 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49579 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P264063 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P33064791 - -
          Agilent Human 1B Microarray G4111A A_32_P437749 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7944529 - -
          Affymetrix Human Gene 1.0 ST Array 7944528 - -
          Affymetrix Human Gene 1.1 ST Array 7944527 - -
          Affymetrix HT HG-U133+ PM Array Plate 1564229_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100009277.hg.1 ENST00000319763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009277.hg.1 ENST00000319763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009277.hg.1 ENST00000319763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009277.hg.1 ENST00000530303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009277.hg.1 ENST00000530303.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100009277.hg.1 ENST00000319763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009277.hg.1 ENST00000319763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009277.hg.1 ENST00000319763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009277.hg.1 ENST00000530303.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100009277.hg.1 ENST00000530303.5 Ensembl
          Human Genome U133 Plus 2.0 Array 1564229_PM_at - -
          Human Genome U133 Plus 2.0 Array 1564229_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564229_at11 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3299424 AK096981.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1810956 AK096981.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0