Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03655764_m1
          See other NTRK3-AS1 GE Assays ›
          Gene Symbol:
          NTRK3-AS1
          Gene Name:
          NTRK3 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 88252730 - 88271066 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03655764_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NTRK3-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038229.1 3 - 4 1057 99
          GenBank mRNA AK097486.1 - 3 - 4 1056 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream neurotrophic receptor tyrosine kinase 3 NTRK3 GP145-TrkC,
          TRKC,
          gp145(trkC)
          -11.1kb Anti-sense Chr. 15 - 87859749 - 88256796 on NCBI Build GRCh38
          Downstream mitochondrial ribosomal protein L46 MRPL46 C15orf4,
          LIECG2,
          P2ECSL
          188.4kb Anti-sense Chr. 15 - 88459478 - 88467402 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500008221.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000836.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815499 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815479 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319511 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16824516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319456 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30319502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09815539 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 147308 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 136618 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 51530 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 116548 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 233291 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 339868 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3853394 - -
          Agilent Human Genome CGH Microarray 44B A_14_P127279 - -
          Agilent Human Genome CGH Microarray 44A A_14_P127279 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 39593 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P127279 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03079929 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3853394 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3853394 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P127279 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03079929 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P127279 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P127279 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 75983 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 25727 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20339226 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12318607 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12320431 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P127279 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03079929 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03079942 - -
          Agilent Human 1B Microarray G4111A A_32_P155571 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P12320431 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P127279 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03079929 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 231469_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR15007692.hg.1 NR_038229.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15007692.hg.1 TC15000836.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008221.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500159190.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500159190.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500159190.hg.1 TC1500008221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500159190.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500159190.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500159192.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500159192.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500159192.hg.1 TC1500008221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500159192.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500159192.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500159191.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500159191.hg.1 NR_038229.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500159191.hg.1 TC1500008221.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500159191.hg.1 ENST00000569588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500159191.hg.1 NR_038229.1 Refseq
          Human Genome U133 Plus 2.0 Array 231469_at - -
          Human Genome U133 Plus 2.0 Array 231469_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231469_at7 - -
          Human Genome U95 Set 81698_at - -
          Human Genome U95 Set 81698_PM_at - -
          Human Genome U133 Set 231469_PM_at - -
          Human Genome U133 Set 231469_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053206 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053206 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0