Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03655835_m1
          See other LOC285804 GE Assays ›
          Gene Symbol:
          LOC285804
          Gene Name:
          uncharacterized LOC285804
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 170266669 - 170276762 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03655835_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC285804
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_126021.1 1 - 2 82 89
          GenBank mRNA AK097676.1 - 1 - 2 82 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB045863.1 - 1 - 2 82 Chr. 6 - 170268063 - 170276762 [-] on Build GRCh38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC154449 LOC154449 -12.8kb Sense Chr. 6 - 170254334 - 170262569 on NCBI Build GRCh38
          Downstream delta like canonical Notch ligand 1 DLL1 DELTA1,
          DL1,
          Delta
          5.4kb Sense Chr. 6 - 170282200 - 170291075 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014030.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 817780 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 354903 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 294581 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 370384 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 217462 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 299152 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 64185 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 288410 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 362544 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 235684 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 331238 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 202844 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 255659 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 369148 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3209135 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 7693 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3209135 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01641686 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 94558 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01641686 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01641678 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15954626 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01641674 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01641686 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196694.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196694.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196694.hg.1 TC0600014030.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196694.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196694.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196690.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196690.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196690.hg.1 TC0600014030.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196690.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196690.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196692.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196692.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196692.hg.1 TC0600014030.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196692.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196692.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196693.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196693.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196693.hg.1 TC0600014030.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196693.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196693.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196691.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196691.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196691.hg.1 TC0600014030.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196691.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196691.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 ENST00000607733.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 NR_126021.1 Refseq
          Affymetrix Human Clariom D Assay TC0600014030.hg.1 NR_126021.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3215138 AK097676.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3232138 - -
          HumanHT-12 V4.0 expression beadchip ILMN_3215138 AK097676.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline