Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03656353_g1
          See other LOC100128079 GE Assays ›
          Gene Symbol:
          LOC100128079
          Gene Name:
          uncharacterized LOC100128079
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 27650134 - 27719029 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03656353_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100128079
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC031310.1 - 1 - 2 101 60
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA256496.1 - 1 - 2 319 Chr. 16 - 27685296 - 27719029 [-] on Build GRCh38 60
          GenBankmRNA BI549724.1 - 1 - 2 101 Chr. 16 - 27708579 - 27718806 [-] on Build GRCh38 60
          GenBankmRNA BM014774.1 - 1 - 2 66 Chr. 16 - 27708445 - 27718776 [-] on Build GRCh38 60
          GenBankmRNA HY118830.1 - 1 - 2 92 Chr. 16 - 27708742 - 27718801 [-] on Build GRCh38 60

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream KIAA0556 KIAA0556 JBTS26 Anti-sense Chr. 16 - 27550128 - 27780371 on NCBI Build GRCh38
          Downstream GSG1 like GSG1L PRO19651 68.7kb Sense Chr. 16 - 27787529 - 28063509 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1569620_PM_s_at - -
          Affymetrix HT HG-U133+ PM Array Plate 243053_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000568831.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000568831.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000568831.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000563052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000564893.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000568831.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000568831.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000568831.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600009840.hg.1 ENST00000610941.4 Ensembl
          Human Genome U133 Plus 2.0 Array 1569620_s_at - -
          Human Genome U133 Plus 2.0 Array 1569620_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 243053_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 243053_x_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243053_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569620_s_at4 - -
          Human Genome U133 Set 243053_x_at - -
          Human Genome U133 Set 243053_PM_x_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline