Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03656456_s1
          See other FGF14-AS2 GE Assays ›
          Gene Symbol:
          FGF14-AS2
          Gene Name:
          FGF14 antisense RNA 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 102394578 - 102395705 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03656456_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FGF14-AS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036487.1 1 601 81
          GenBank mRNA BC033993.1 - 1 550 81
          BX096285.1 - 1 601 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300007883.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000366.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P79483 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17661 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21822 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P79483 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P79483 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P79483 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1486479 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1486479 - -
          Agilent Human 1B Microarray G4111A A_32_P79481 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 243225_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 230351_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1300007883.hg.1 ENST00000606448.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300007883.hg.1 NR_036487.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300007883.hg.1 NR_036487.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13003917.hg.1 NR_036487.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13003917.hg.1 TC13000366.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1300153026.hg.1 NR_036487.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300153026.hg.1 TC1300007883.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300153026.hg.1 NR_036487.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300153027.hg.1 ENST00000606448.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300153027.hg.1 NR_036487.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300153027.hg.1 TC1300007883.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300153027.hg.1 ENST00000606448.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300153027.hg.1 NR_036487.1 Refseq
          Affymetrix Human Clariom D Assay TC1300007883.hg.1 ENST00000606448.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300007883.hg.1 NR_036487.1 Refseq
          Affymetrix Human Clariom D Assay TC1300007883.hg.1 NR_036487.1 Refseq
          Human Genome U133 Plus 2.0 Array 230351_at - -
          Human Genome U133 Plus 2.0 Array 243225_at - -
          Human Genome U133 Plus 2.0 Array 230351_PM_at - -
          Human Genome U133 Plus 2.0 Array 243225_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230351_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243225_at4 - -
          Human Genome U95 Set 53170_at - -
          Human Genome U95 Set 53170_PM_at - -
          Human Genome U95 Set 90800_PM_r_at - -
          Human Genome U95 Set 90800_r_at - -
          Human Genome U133 Set 243225_PM_at - -
          Human Genome U133 Set 243225_at - -
          Human Genome U133 Set 230351_PM_at - -
          Human Genome U133 Set 230351_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050440 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050440 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3274671 NR_036487.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3274671 AF086059.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3274671 AF339777.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3274671 BC033993.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3274671 BM674599.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0