Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03656574_m1
          See other LOC400620 GE Assays ›
          Gene Symbol:
          LOC400620
          Gene Name:
          uncharacterized LOC400620
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 73786822 - 73800795 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03656574_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC400620
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040020.1 6 - 7 2113 106
          GenBank mRNA BC035399.1 - 6 - 7 2113 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100134391 LOC100134391 -42.4kb Sense Chr. 17 - 73737854 - 73757723 on NCBI Build GRCh38
          Downstream ribosomal protein L38 RPL38 L38 402.9kb Sense Chr. 17 - 74203656 - 74209880 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008835.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000824.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458959 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458966 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042341 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042343 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042326 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17014846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042379 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042360 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042377 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042339 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042462 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042405 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042356 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31042457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10458848 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P85348 - -
          Agilent Human 1B Microarray G4111A A_32_P416840 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1557731_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1562000_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000584660.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 ENST00000584660.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010612.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010612.hg.1 NR_040020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010612.hg.1 TC17000824.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000584660.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 ENST00000584660.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay TC1700008835.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700165913.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700165913.hg.1 NR_040020.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700165913.hg.1 TC1700008835.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700165913.hg.1 ENST00000455366.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700165913.hg.1 NR_040020.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557731_PM_at - -
          Human Genome U133 Plus 2.0 Array 1562000_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557731_at - -
          Human Genome U133 Plus 2.0 Array 1562000_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557731_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562000_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009896 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009896 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline