Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03656851_s1
          See other LOC729040 GE Assays ›
          Gene Symbol:
          LOC729040
          Gene Name:
          uncharacterized LOC729040
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 91279964 - 91281136 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03656851_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC729040
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF452720.1 - 1 253 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CB104807.1 - 1 253 Chr. 5 - 91280488 - 91281142 [-] on Build GRCh38 84
          GenBankmRNA CB108865.1 - 1 253 Chr. 5 - 91280758 - 91281142 [-] on Build GRCh38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream adhesion G protein-coupled receptor V1 ADGRV1 FEB4,
          GPR98,
          MASS1,
          USH2B,
          USH2C,
          VLGR1,
          VLGR1b
          -115.9kb Anti-sense Chr. 5 - 90558681 - 91164226 on NCBI Build GRCh38
          Downstream lung cancer associated transcript 1 (non-protein coding) LUCAT1 SCAL1 21.8kb Sense Chr. 5 - 91302986 - 91314402 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF452720.1 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15719955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097749 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097740 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04097731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24277545 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 249338 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 66034 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 1996 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 122791 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 282505 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 34894 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3414107 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17208842 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8113071 - -
          Affymetrix Human Transcriptome Array 2.0 TC0500011445.hg.1 ENST00000440769.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500011445.hg.1 ENST00000440769.3 Ensembl
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1872261 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 1110309 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_95916 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0