Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03656915_m1
          See other BLACE GE Assays ›
          Gene Symbol:
          BLACE
          Gene Name:
          B-cell acute lymphoblastic leukemia expressed
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 155356988 - 155367933 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03656915_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BLACE
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_103545.1 5 - 6 666 67
          GenBank mRNA AY166699.1 - 5 - 6 675 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream insulin induced gene 1 INSIG1 CL-6,
          CL6
          -46.8kb Anti-sense Chr. 7 - 155297776 - 155310235 on NCBI Build GRCh38
          Downstream engrailed homeobox 2 EN2 98.1kb Anti-sense Chr. 7 - 155458129 - 155464831 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013178.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16044762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887119 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887128 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16044797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16044790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887254 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16044796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16044774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156082 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887171 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16044793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26155981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26155977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26155990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05887162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26156002 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3405023 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14438 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01841024 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 110019 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01841027 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01841024 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P25860253 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01841018 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38265197 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195084.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195084.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195084.hg.1 TC0700013178.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700195084.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195084.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195083.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195083.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195083.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195083.hg.1 TC0700013178.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700195083.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195083.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195083.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195081.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195081.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195081.hg.1 TC0700013178.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700195081.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195081.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013178.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195080.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195080.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195080.hg.1 TC0700013178.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700195080.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195080.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195082.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195082.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195082.hg.1 NR_103545.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700195082.hg.1 TC0700013178.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700195082.hg.1 ENST00000378120.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195082.hg.1 ENST00000616210.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700195082.hg.1 NR_103545.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline