Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657225_s1
          See other LOC286121 GE Assays ›
          Gene Symbol:
          LOC286121
          Gene Name:
          uncharacterized LOC286121
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657225_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC286121
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC039302.1 - 1 3255 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU736510.1 - 1 Chr. 8 - 142406618 - 142407008 [-] on Build GRCh38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream t-SNARE domain containing 1 TSNARE1 -361.0 Anti-sense Chr. 8 - 142212080 - 142403291 on NCBI Build GRCh38
          Downstream adhesion G protein-coupled receptor B1 ADGRB1 BAI1,
          GDAIF
          42.9kb Sense Chr. 8 - 142449703 - 142545009 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26812542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26812537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06528881 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 890249 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 280273 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 201060 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 40898 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 236390 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 81997 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129387 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 250101 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 218545 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3463355 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 24212 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3463355 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16706365 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561671_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0800009142.hg.1 ENST00000569285.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009142.hg.1 ENST00000569285.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1561671_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561671_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561671_at2 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0