Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657363_g1
          See other LOC400748 GE Assays ›
          Gene Symbol:
          LOC400748
          Gene Name:
          uncharacterized LOC400748
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 30858158 - 30862399 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657363_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC400748
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC040627.1 - 1 - 2 213 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM908199.1 - 224 Chr. - - [-] on Build 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream lysosomal protein transmembrane 5 LAPTM5 CLAST6 -100.3kb Anti-sense Chr. 1 - 30732468 - 30757840 on NCBI Build GRCh38
          Downstream syndecan 3 SDC3 SDCN,
          SYND3
          9.2kb Anti-sense Chr. 1 - 30869466 - 30909735 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139339 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139393 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15045111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139355 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139357 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139341 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139338 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139381 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116765 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139350 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116761 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116800 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00116769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20139368 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3714325 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55719 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3714325 - -
          Agilent Human 1B Microarray G4111A A_32_P920289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1564401_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100007620.hg.1 ENST00000431202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007620.hg.1 ENST00000431202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007620.hg.1 ENST00000431202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007620.hg.1 ENST00000431202.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1564401_PM_at - -
          Human Genome U133 Plus 2.0 Array 1564401_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564401_at5 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0