Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657693_m1
          See other DNAJC9-AS1 GE Assays ›
          Gene Symbol:
          DNAJC9-AS1
          Gene Name:
          DNAJC9 antisense RNA 1
          Gene Aliases:
          C10orf103, bA537A6.3
          Chromosome Location:
          Chr.10: 73247367 - 73276984 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657693_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DNAJC9-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038373.1 2 - 3 175 76
          GenBank mRNA BC043233.1 - 2 - 3 183 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI560079.1 - 2 - 3 183 Chr. 10 - 73247360 - 73274941 [+] on Build GRCh38 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cilia and flagella associated protein 70 CFAP70 TTC18 Anti-sense Chr. 10 - 73253758 - 73361064 on NCBI Build GRCh38
          Downstream annexin A7 ANXA7 ANX7,
          SNX,
          SYNEXIN
          102.5kb Anti-sense Chr. 10 - 73375101 - 73414085 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008022.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000459.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human 1B Microarray G4111A A_32_P343829 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1557827_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR10005420.hg.1 ENST00000413597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005420.hg.1 ENST00000440197.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005420.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005420.hg.1 TC10000459.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000513954.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 ENST00000513954.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005417.hg.1 ENST00000440197.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005417.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005417.hg.1 TC10000459.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10005421.hg.1 ENST00000413597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005421.hg.1 ENST00000440197.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005421.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005421.hg.1 TC10000459.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10005422.hg.1 ENST00000440197.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005422.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005422.hg.1 TC10000459.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10005418.hg.1 ENST00000440197.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10005418.hg.1 NR_038373.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10005418.hg.1 TC10000459.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000154379.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154379.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154379.hg.1 TC1000008022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154379.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154379.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154375.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154375.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154375.hg.1 TC1000008022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154375.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154375.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154377.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154377.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154377.hg.1 TC1000008022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154377.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154377.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154372.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154372.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154372.hg.1 TC1000008022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154372.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154372.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154373.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154373.hg.1 ENST00000513954.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154373.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154373.hg.1 TC1000008022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154373.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154373.hg.1 ENST00000513954.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154373.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154378.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154378.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154378.hg.1 TC1000008022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154378.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154378.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000513954.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 ENST00000513954.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008022.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154376.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154376.hg.1 NR_038373.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000154376.hg.1 TC1000008022.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000154376.hg.1 ENST00000440197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000154376.hg.1 NR_038373.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557827_at - -
          Human Genome U133 Plus 2.0 Array 1557827_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557827_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044110 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044110 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0