Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011421.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011421.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011421.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011421.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011421.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011421.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188083.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188083.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188083.hg.1 |
TC1100011421.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100188083.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188083.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188084.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188084.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100188084.hg.1 |
TC1100011421.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100188084.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100188084.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011421.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011421.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011421.hg.1 |
ENST00000562772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011421.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011421.hg.1 |
NR_104161.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011421.hg.1 |
NR_104161.1 |
Refseq |