Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657807_m1
          See other LOC339192 GE Assays ›
          Gene Symbol:
          LOC339192
          Gene Name:
          uncharacterized LOC339192
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 45219313 - 45219974 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03657807_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC339192
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC045769.1 - 3 - 4 658 58
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG490770.1 - 1 - 2 166 Chr. 17 - 45218674 - 45220425 [-] on Build GRCh38 58

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105371795 LOC105371795 -46.2kb Sense Chr. 17 - 45168805 - 45171485 on NCBI Build GRCh38
          Downstream formin like 1 FMNL1 C17orf1,
          C17orf1B,
          FHOD4,
          FMNL,
          KW-13
          1.5kb Anti-sense Chr. 17 - 45221728 - 45247322 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30911384 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30911387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10337733 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10337729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10337734 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30911380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10337737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30911383 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 86851 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 432113 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 96134 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 690710 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 23521 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03250014 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03250014 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20656450 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P20875372 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20656447 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20656444 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03250014 - -
          Agilent Human 1B Microarray G4111A A_32_P921659 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P20656447 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03250014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000590522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000590522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000585471.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000586376.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000590495.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000590522.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000590522.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010872.hg.1 ENST00000591365.1 Ensembl
          Human Genome U95 Set 46078_PM_at - -
          Human Genome U95 Set 51390_PM_at - -
          Human Genome U95 Set 51390_at - -
          Human Genome U95 Set 46078_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3289508 CR604709.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline