Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03657957_s1
          See other LOC646778 GE Assays ›
          Gene Symbol:
          LOC646778 B4GALT6
          Gene Name:
          uncharacterized LOC646778
          beta-1,4-galactosyltransferase 6
          Gene Aliases:
          -
          B4Gal-T6, beta4Gal-T6
          Chromosome Location:
          -
          Chromosome Location:
          Chr.18: 31622246 - 31724670 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03657957_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC646778
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC054864.1 - 1 550 98
          Gene Symbol: B4GALT6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017026089.1 XP_016881578.1 1 98
          XM_017026092.1 XP_016881581.1 1 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX099294.1 - 1 Chr. 18 - 31685875 - 31686627 [+] on Build GRCh38 98
          GenBankmRNA BU177880.1 - 1 Chr. 18 - 31686035 - 31686867 [+] on Build GRCh38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transthyretin TTR CTS,
          CTS1,
          HEL111,
          HsT2651,
          PALB,
          TBPA
          -86.6kb Sense Chr. 18 - 31591767 - 31599024 on NCBI Build GRCh38
          Downstream leucine rich repeat containing 37 member A7,
          pseudogene
          LRRC37A7P 37.4kb Sense Chr. 18 - 31724227 - 31725686 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the XM_017026089.1, XM_017026092.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          keratan sulfate biosynthetic process
          beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase activity
          galactosyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10610410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10610419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10610407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31210097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10610413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10610411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10610409 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31210107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10610421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31210098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31210106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31210108 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 377642 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610421 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610407 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210097 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210100 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210107 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210108 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610410 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610413 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610409 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610419 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610420 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610411 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210098 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210106 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210096 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610409 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610419 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210108 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610413 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210107 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610421 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210100 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210097 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210106 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610411 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610420 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610410 - -
          Agilent Whole Chr array_18 19 21 A_17_P10610407 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210096 - -
          Agilent Whole Chr array_18 19 21 A_17_P31210098 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03337779 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 243452_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1800007019.hg.1 ENST00000565978.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007019.hg.1 ENST00000565978.1 Ensembl
          Human Genome U133 Plus 2.0 Array 231342_at - -
          Human Genome U133 Plus 2.0 Array 243452_PM_at - -
          Human Genome U133 Plus 2.0 Array 231342_PM_at - -
          Human Genome U133 Plus 2.0 Array 243452_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 243452_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231342_at9 - -
          Human Genome U95 Set 57475_at - -
          Human Genome U95 Set 57475_PM_at - -
          Human Genome U95 Set 72671_PM_r_at - -
          Human Genome U95 Set 72671_r_at - -
          Human Genome U133 Set 231342_PM_at - -
          Human Genome U133 Set 243452_PM_at - -
          Human Genome U133 Set 243452_at - -
          Human Genome U133 Set 231342_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1796064 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1678524 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1678524 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1796064 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1796064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6cbc4fbdf-xtbd4:80/100.66.128.131:80.
          git-commit: 26a732473c29a7560faabcf99da408bd94702681
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc