Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03659511_s1
          See other LOC100130157 GE Assays ›
          Gene Symbol:
          LOC100130157
          Gene Name:
          uncharacterized LOC100130157
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03659511_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100130157
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK130385.1 - 1 1251 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphatidylinositol glycan anchor biosynthesis class T PIGT CGI-06,
          MCAHS3,
          NDAP,
          PNH2
          -19.2kb Sense Chr. 20 - 45416067 - 45426245 on NCBI Build GRCh38
          Downstream WAP four-disulfide core domain 2 WFDC2 EDDM4,
          HE4,
          WAP5,
          dJ461P17.6
          68.0 Sense Chr. 20 - 45469754 - 45481532 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835323 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17194881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835366 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17194883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835321 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835318 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31835309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11187485 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59574 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3270136 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000424705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000424705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000424705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000424705.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000424705.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000424705.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007490.hg.1 ENST00000595203.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3272595 AK130385.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0