Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03659876_m1
          See other LOC388406 GE Assays ›
          Gene Symbol:
          LOC388406
          Gene Name:
          uncharacterized LOC388406
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 60564546 - 60586620 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03659876_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC388406
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_126009.1 3 - 4 330 74
          GenBank mRNA AK131012.1 - 3 - 4 331 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI125233.1 - 1 - 2 Chr. 17 - 60565377 - 60586623 [-] on Build GRCh38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream amyloid beta precursor protein binding protein 2 APPBP2 APP-BP2,
          HS.84084,
          PAT1
          -51.0kb Anti-sense Chr. 17 - 60443149 - 60526240 on NCBI Build GRCh38
          Downstream protein phosphatase,
          Mg2+/Mn2+ dependent 1D
          PPM1D PP2C-DELTA,
          WIP1
          13.6kb Sense Chr. 17 - 60600183 - 60666280 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401120 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980810 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401119 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17002576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980831 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980836 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17002568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401112 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17002578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10401134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17002573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30980869 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 57968 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 59901 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 446055 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 527632 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 256061 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 113051 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 380755 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 239696 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 384995 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 473662 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3748769 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 5694 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P20693316 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3748769 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03270347 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03270347 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P20693331 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 58373 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 72502 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03270347 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20693331 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20693314 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20693316 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03270340 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03270333 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40866183 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20693357 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20693341 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03270347 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P20693331 - -
          Array Name Probe IDs Targets Target Type
          Human Genome U95 Set 87399_at - -
          Human Genome U95 Set 87399_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc