Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03659945_s1
          See other LOC645769 GE Assays ›
          Gene Symbol:
          LOC645769
          Gene Name:
          uncharacterized LOC645769
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03659945_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC645769
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK123791.1 - 1 1152 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transcription elongation factor A N-terminal and central domain containing TCEANC Anti-sense Chr. X - 13653106 - 13665409 on NCBI Build GRCh38
          Downstream RAB9A,
          member RAS oncogene family
          RAB9A RAB9 35.7kb Anti-sense Chr. X - 13689121 - 13709825 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 573985 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3418992 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28608 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3418992 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P41591580 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41591580 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P41591580 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_040624_at - -
          Affymetrix Human Gene 1.0 ST Array XR_015325_at - -
          Affymetrix Human Gene 1.0 ST Array XR_015458_at - -
          Affymetrix Human Gene 1.0 ST Array 8171351 - -
          Affymetrix Human Gene 1.1 ST Array 8171350 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1719028 AK123791.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1660873 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1719028 AK123791.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1660873 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1719028 AK123791.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1660873 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1719028 AK123791.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1719028 AK123791.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1660873 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1719028 AK123791.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1660873 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1719028 AK123791.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0