Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035999.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035999.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035999.hg.1 |
TC02002216.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201998.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201998.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201998.hg.1 |
TC0200013909.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200201998.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201998.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
ENST00000457336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013909.hg.1 |
NR_033871.1 |
Refseq |