Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03661754_s1
          See other LOC100129581 GE Assays ›
          Gene Symbol:
          LOC100129581
          Gene Name:
          uncharacterized LOC100129581
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03661754_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129581
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK125905.1 - 1 3452 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB340012.1 - 1 Chr. 2 - 7847353 - 7847923 [+] on Build GRCh38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100506274 LOC100506274 -397.1kb Anti-sense Chr. 2 - 7421261 - 7450254 on NCBI Build GRCh38
          Downstream uncharacterized LOC101929551 LOC101929551 35.5kb Sense Chr. 2 - 7886767 - 7899655 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15214625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972350 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972320 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15214629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15214631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049615 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972381 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972328 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972325 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972347 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972360 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972366 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972336 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049634 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972343 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00972333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21049635 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 808994 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3258076 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 31541 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13098588 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001718835_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001726573_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001721419_at - -
          Affymetrix Human Gene 1.0 ST Array 8050114 - -
          Affymetrix Human Gene 1.1 ST Array 8050113 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0