Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03662299_s1
          See other LOC100129840 GE Assays ›
          Gene Symbol:
          LOC100129840
          Gene Name:
          uncharacterized LOC100129840
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03662299_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129840
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK126633.1 - 1 2949 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CD298819.1 - 218 Chr. - - [-] on Build 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream integrin subunit alpha 11 ITGA11 HsT18964 -138.7kb Sense Chr. 15 - 68297433 - 68432312 on NCBI Build GRCh38
          Downstream acidic nuclear phosphoprotein 32 family member A ANP32A C15orf1,
          HPPCn,
          I1PP2A,
          LANP,
          MAPM,
          PHAP1,
          PHAPI,
          PP32
          203.8kb Sense Chr. 15 - 68778535 - 68820922 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733765 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16800792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226981 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09733789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30226994 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 413919 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3305088 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42577 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P33737714 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P33737714 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3305088 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03053958 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 80151 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03053958 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03053961 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40442832 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03053958 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix