Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03662411_s1
          See other LOC100128498 GE Assays ›
          Gene Symbol:
          LOC100128498
          Gene Name:
          uncharacterized LOC100128498
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03662411_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100128498
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK126742.1 - 1 2052 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ZFP90 zinc finger protein ZFP90 FIK,
          NK10,
          ZNF756,
          zfp-90
          -15.3kb Sense Chr. 16 - 68533484 - 68576072 on NCBI Build GRCh38
          Downstream cadherin 3 CDH3 CDHP,
          HJMD,
          PCAD
          49.8kb Sense Chr. 16 - 68644820 - 68727573 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10090024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30629888 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 294374 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3394694 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 25172 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3394694 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P33954951 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P33954951 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300010122.hg.1 ENST00000629101.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007310.hg.1 ENST00000577585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008202.hg.1 ENST00000612545.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010122.hg.1 ENST00000629101.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008202.hg.1 ENST00000612545.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007310.hg.1 ENST00000577585.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3256184 AK126742.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3256184 AK126742.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0