Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03664390_s1
          See other LOC100131719 GE Assays ›
          Gene Symbol:
          LOC100131719
          Gene Name:
          uncharacterized LOC100131719
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03664390_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100131719
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK128441.1 - 1 2455 107
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB301278.1 - 1 Chr. 10 - 123192655 - 123193175 [-] on Build GRCh38 107
          GenBankmRNA DB255023.1 - 1 223 Chr. 10 - 123192589 - 123193159 [+] on Build GRCh38 107
          GenBankmRNA AA137016.1 - 1 186 Chr. 10 - 123192625 - 123193060 [+] on Build GRCh38 107

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream BUB3,
          mitotic checkpoint protein
          BUB3 BUB3L,
          hBUB3
          -25.0kb Sense Chr. 10 - 123154244 - 123165370 on NCBI Build GRCh38
          Downstream G protein-coupled receptor 26 GPR26 473.2kb Sense Chr. 10 - 123666355 - 123697399 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16392436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539634 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930399 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16392425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930408 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16392429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930405 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930401 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930403 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930423 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16392433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930456 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07539595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930400 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27930467 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 487053 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 675851 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375413 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58895 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19096927 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3375413 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 101617 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19096927 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02362298 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0