Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03664641_s1
          See other LOC100133857 GE Assays ›
          Gene Symbol:
          LOC100133857
          Gene Name:
          uncharacterized LOC100133857
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03664641_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100133857
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK128800.1 - 1 1750 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 2 member 10 SLC2A10 ATS,
          GLUT10
          -12.6kb Anti-sense Chr. 20 - 46708358 - 46736347 on NCBI Build GRCh38
          Downstream EYA transcriptional coactivator and phosphatase 2 EYA2 EAB1 143.7kb Anti-sense Chr. 20 - 46894624 - 47188844 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11192420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31840562 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 294824 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3280360 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49466 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03525633 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3280360 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0