Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03666370_m1
          See other CYP51A1-AS1 GE Assays ›
          Gene Symbol:
          CYP51A1-AS1
          Gene Name:
          CYP51A1 antisense RNA 1
          Gene Aliases:
          LRRD1-AS1
          Chromosome Location:
          Chr.7: 92134563 - 92180725 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03666370_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYP51A1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_122109.1 4 - 5 1122 103
          NR_122110.1 2 - 3 604 103
          GenBank mRNA AL133568.1 - 4 - 5 1081 103
          HY053824.1 - 2 - 3 285 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leucine rich repeats and death domain containing 1 LRRD1 Anti-sense Chr. 7 - 92144884 - 92165276 on NCBI Build GRCh38
          Downstream KRIT1,
          ankyrin repeat containing
          KRIT1 CAM,
          CCM1
          19.1kb Anti-sense Chr. 7 - 92198969 - 92246128 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700008319.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05611843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862906 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05611848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862921 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862916 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05611850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862907 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05611858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05611854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25863008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05611948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25862895 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 25037 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 287758 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3873533 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12977 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 17195 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3873533 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3873533 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P171984 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18025024 - -
          Agilent Human 1B Microarray G4111A A_32_P480782 - -
          Agilent Human 1B Microarray G4111A A_32_P413220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 233942_PM_at - -
          Affymetrix Human Genome U219 Array 11761153_PM_s_at - -
          Affymetrix Human Genome U219 Array 11761153_s_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158045.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158045.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158045.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158045.hg.1 TC0700008319.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700158045.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158045.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158045.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000343318.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000430130.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000437357.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 ENST00000458448.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013585.hg.1 NM_001161528.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158044.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158044.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158044.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158044.hg.1 TC0700008319.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700158044.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158044.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158044.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008319.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158046.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158046.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158046.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158046.hg.1 TC0700008319.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700158046.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158046.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158046.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158047.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158047.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158047.hg.1 NR_122110.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158047.hg.1 TC0700008319.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700158047.hg.1 ENST00000453068.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158047.hg.1 NR_122109.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700158047.hg.1 NR_122110.1 Refseq
          Human Genome U133 Plus 2.0 Array 233942_at - -
          Human Genome U133 Plus 2.0 Array 233942_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233942_at6 - -
          Human Genome U95 Set 84184_at - -
          Human Genome U95 Set 84184_PM_at - -
          Human Genome U133 Set 233942_at - -
          Human Genome U133 Set 233942_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0