Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03666530_m1
          See other EXTL3-AS1 GE Assays ›
          Gene Symbol:
          EXTL3-AS1
          Gene Name:
          EXTL3 antisense RNA 1
          Gene Aliases:
          C8orf50
          Chromosome Location:
          Chr.8: 28698403 - 28701464 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03666530_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EXTL3-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_126027.1 2 - 3 709 101
          GenBank mRNA BC118569.1 - 2 - 3 709 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream frizzled class receptor 3 FZD3 Fz-3 -124.1kb Anti-sense Chr. 8 - 28494205 - 28574268 on NCBI Build GRCh38
          Downstream exostosin like glycosyltransferase 3 EXTL3 BOTV,
          EXTL1L,
          EXTR1,
          REGR,
          RPR
          1.4kb Anti-sense Chr. 8 - 28701473 - 28753690 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800009987.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06034631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06034623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06034616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06034609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06034622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06034611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06034615 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26316887 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 166193 AF337817.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 166193 AK131266.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 166193 NM_014491.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 166193 NM_148898.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 166193 NM_148899.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 733967 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3570974 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 11665 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3570974 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01886927 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01886932 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556436_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800167935.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167935.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800167935.hg.1 TC0800009987.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800167935.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167935.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800167934.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167934.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800167934.hg.1 TC0800009987.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800167934.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167934.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000519870.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay TC0800009987.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 NR_126027.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 TC0800009987.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000501224.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000520023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 ENST00000523789.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800167931.hg.1 NR_126027.1 Refseq
          Human Genome U133 Plus 2.0 Array 1556436_at - -
          Human Genome U133 Plus 2.0 Array 1556436_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556436_at8 - -
          Human Genome U95 Set 80243_PM_at - -
          Human Genome U95 Set 80243_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0