Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03666840_s1
          See other DGCR10 GE Assays ›
          Gene Symbol:
          DGCR10
          Gene Name:
          DiGeorge syndrome critical region gene 10 (non-protein coding)
          Gene Aliases:
          DGS-B
          Chromosome Location:
          Chr.22: 19022624 - 19023550 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03666840_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DGCR10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026651.1 1 511 113
          GenBank mRNA BM681417.1 - 1 113
          L77559.1 - 1 350 113
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM727772.1 - 1 70 Chr. 22 - 19023065 - 19023494 [+] on Build GRCh38 113
          GenBankmRNA DB298165.1 - 1 Chr. 22 - 19023019 - 19023538 [-] on Build GRCh38 113

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream DiGeorge syndrome critical region gene 5 (non-protein coding) DGCR5 LINC00037,
          NCRNA00037
          Sense Chr. 22 - 18970498 - 19031242 on NCBI Build GRCh38
          Downstream DiGeorge syndrome critical region gene 2 DGCR2 DGS-C,
          IDD,
          LAN,
          SEZ-12
          13.1kb Anti-sense Chr. 22 - 19036282 - 19122454 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200006580.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000038.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32097771 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 88915 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 31466 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 407074 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3362267 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4694 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4695 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14246 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03600450 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03600450 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3362267 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3362267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32097771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32097771 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1655044 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1655044 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11427695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254409 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11427703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11427699 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11427700 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11427708 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11427698 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254423 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11427705 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P22613412 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03600450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1557451_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200006580.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006580.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006580.hg.1 NR_026651.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006580.hg.1 NR_026651.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006580.hg.1 NR_026651.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006580.hg.1 NR_026651.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22000344.hg.1 TC22000038.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22000344.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145092.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145092.hg.1 TC2200006580.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200145092.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145091.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145091.hg.1 TC2200006580.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200145091.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145093.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200145093.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145093.hg.1 TC2200006580.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200145093.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200145093.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145094.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200145094.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200145094.hg.1 TC2200006580.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200145094.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200145094.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006580.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006580.hg.1 ENST00000609839.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006580.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006580.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006580.hg.1 NR_026651.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006580.hg.1 NR_026651.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557451_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557451_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557451_at3 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A020071 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A020071 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3311025 NR_026651.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3311025 NR_026651.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3311025 NR_026651.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3311025 NR_026651.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0