Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03667163_s1
          See other HPYR1 GE Assays ›
          Gene Symbol:
          HPYR1
          Gene Name:
          Helicobacter pylori responsive 1 (non-protein coding)
          Gene Aliases:
          HPRG1, LINC00027, NCRNA00027
          Chromosome Location:
          Chr.8: 132560498 - 132561479 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03667163_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HPYR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026684.1 1 655 99
          GenBank mRNA AF200341.1 - 1 655 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream potassium voltage-gated channel subfamily Q member 3 KCNQ3 BFNC2,
          EBN2,
          KV7.3
          -79.7kb Sense Chr. 8 - 132120858 - 132480757 on NCBI Build GRCh38
          Downstream leucine rich repeat containing 6 LRRC6 CILD19,
          LRTP,
          TSLRP
          8.9kb Sense Chr. 8 - 132570419 - 132685039 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800011860.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08001653.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482356 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764658 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482357 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482359 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482322 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482360 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16136670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482326 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482323 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482324 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26764690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06482340 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3311825 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4719 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4720 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19979 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3311825 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3311825 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482393 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482388 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482394 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482399 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482393 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482388 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482394 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482399 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26764709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06482353 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482335 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482362 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482319 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482333 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482360 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482350 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482338 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482371 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482352 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482328 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16136674 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16136671 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482369 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482383 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16136670 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482317 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482330 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482324 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482392 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482386 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482394 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482358 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482331 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482388 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482373 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482380 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06482391 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1566916_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1566917_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR08021469.hg.1 ENST00000520457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08021469.hg.1 NR_026684.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08021469.hg.1 TC08001653.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800011860.hg.1 ENST00000520457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011860.hg.1 NR_026684.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800180546.hg.1 ENST00000520457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800180546.hg.1 NR_026684.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800180546.hg.1 TC0800011860.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800180546.hg.1 ENST00000520457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800180546.hg.1 NR_026684.1 Refseq
          Affymetrix Human Clariom D Assay TC0800011860.hg.1 ENST00000520457.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011860.hg.1 NR_026684.1 Refseq
          Human Genome U133 Plus 2.0 Array 1566917_PM_at - -
          Human Genome U133 Plus 2.0 Array 1566916_PM_at - -
          Human Genome U133 Plus 2.0 Array 1566916_at - -
          Human Genome U133 Plus 2.0 Array 1566917_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566916_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566917_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036148 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036148 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309940 NR_026684.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3309940 AF200341.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309940 NR_026684.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309940 AF200341.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309940 NR_026684.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309940 AF200341.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309940 NR_026684.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309940 AF200341.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0