Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03667613_s1
          See other LOC340085 GE Assays ›
          Gene Symbol:
          LOC340085
          Gene Name:
          uncharacterized LOC340085
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03667613_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC340085
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK021736.1 - 1 647 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AL705126.1 - 1 Chr. 5 - 57264348 - 57265036 [-] on Build GRCh38 97
          GenBankmRNA AL705128.1 - 1 Chr. 5 - 57264301 - 57265036 [-] on Build GRCh38 97
          GenBankmRNA AU118965.1 - 1 647 Chr. 5 - 57264253 - 57265078 [+] on Build GRCh38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream MIER family member 3 MIER3 -312.1kb Anti-sense Chr. 5 - 56919602 - 56952130 on NCBI Build GRCh38
          Downstream actin,
          beta like 2
          ACTBL2 ACT 214.2kb Anti-sense Chr. 5 - 57480016 - 57482809 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 124662 AF428253.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 124662 AK095506.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 124662 BC034360.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 124662 NM_206808.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 8x60K 32992 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 291003 - -
          Agilent Human Genome CGH Microarray 44B A_14_P139114 - -
          Agilent Human Genome CGH Microarray 44A A_14_P139114 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P139114 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P139114 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P139114 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 48386 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 96665 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P139114 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P139114 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 232455_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 232455_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 232455_x_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232455_x_at9 - -
          Human Genome U133 Set 232455_PM_x_at - -
          Human Genome U133 Set 232455_x_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0