Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03668047_s1
          See other LOC90246 GE Assays ›
          Gene Symbol:
          LOC90246
          Gene Name:
          uncharacterized LOC90246
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 128507835 - 128510595 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03668047_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC90246
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026954.1 1 2160 86
          GenBank mRNA AK023635.1 - 1 1852 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream GATA2 antisense RNA 1 GATA2-AS1 -4.9kb Sense Chr. 3 - 128489203 - 128503207 on NCBI Build GRCh38
          Downstream long intergenic non-protein coding RNA 1565 LINC01565 C3orf27,
          GR6
          61.4kb Anti-sense Chr. 3 - 128572000 - 128576086 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300008725.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000676.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15487073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22714587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22714559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15487065 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574992 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15487071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22714573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22714571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02574989 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 259594 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 344197 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P532180 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P532180 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 38868 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 54905 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P532180 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P532180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574970 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574966 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714546 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574967 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714559 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15487065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574965 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714543 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574971 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574977 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574975 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574968 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574965 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714546 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574975 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574971 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714559 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574977 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15487065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574966 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574970 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714542 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574967 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22714543 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02574969 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P532180 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P532180 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00808662 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574988 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574998 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574952 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15487072 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574996 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574971 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574980 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15487071 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574963 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15487056 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574965 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574975 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574960 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574994 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574957 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574970 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574968 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15487063 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574984 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574986 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574959 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574973 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574958 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15487064 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02574992 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 233835_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 233830_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR03012114.hg.1 NR_026954.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03012114.hg.1 TC03000676.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300162042.hg.1 NR_026954.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300162042.hg.1 TC0300008725.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300162042.hg.1 NR_026954.1 Refseq
          Human Genome U133 Plus 2.0 Array 233835_at - -
          Human Genome U133 Plus 2.0 Array 233830_PM_at - -
          Human Genome U133 Plus 2.0 Array 233830_at - -
          Human Genome U133 Plus 2.0 Array 233835_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233835_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233830_at7 - -
          Human Genome U95 Set 92157_PM_r_at - -
          Human Genome U95 Set 92157_r_at - -
          Human Genome U133 Set 233830_at - -
          Human Genome U133 Set 233835_at - -
          Human Genome U133 Set 233830_PM_at - -
          Human Genome U133 Set 233835_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023164 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0