Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03669270_s1
          See other ARPIN GE Assays ›
          Gene Symbol:
          ARPIN
          Gene Name:
          actin-related protein 2/3 complex inhibitor
          Gene Aliases:
          C15orf38
          Chromosome Location:
          Chr.15: 89896532 - 89912990 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03669270_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARPIN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282380.1 NP_001269309.1 6 4809 78
          NM_182616.3 NP_872422.1 6 4420 78
          GenBank mRNA AK055252.1 - 1 847 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF673840.1 - 157 Chr. - - [-] on Build 78
          GenBankmRNA CN274423.1 - 1 274 Chr. 15 - 89897860 - 89898480 [-] on Build GRCh38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream C15orf38-AP3S2 readthrough C15orf38-AP3S2 Sense Chr. 15 - 89830599 - 89912990 on NCBI Build GRCh38
          Downstream zinc finger protein 710 ZNF710 100.0kb Anti-sense Chr. 15 - 90001491 - 90082200 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ARPIN

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell migration
          directional locomotion
          negative regulation of actin nucleation
          negative regulation of lamellipodium morphogenesis
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P196384 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P196384 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 16623 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 41897 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P196384 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P196384 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P196384 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 228791_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0300006995.hg.1 ENST00000516891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007954.hg.1 ENST00000630657.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008775.hg.1 ENST00000515903.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008281.hg.1 ENST00000516836.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011940.hg.1 ENST00000607576.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007954.hg.1 ENST00000630657.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008281.hg.1 ENST00000516836.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 ENST00000504333.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010838.hg.1 NR_109904.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011940.hg.1 ENST00000607576.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006995.hg.1 ENST00000516891.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008775.hg.1 ENST00000515903.2 Ensembl
          Human Genome U133 Plus 2.0 Array 217548_at - -
          Human Genome U133 Plus 2.0 Array 228791_PM_at - -
          Human Genome U133 Plus 2.0 Array 228791_at - -
          Human Genome U133 Plus 2.0 Array 217548_PM_at - -
          Human Genome U133A 2.0 Array 217548_at - -
          Human Genome U133A 2.0 Array 217548_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228791_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217548_at8 - -
          Human Genome U95 Set 53726_r_at - -
          Human Genome U95 Set 53726_PM_r_at - -
          Human Genome U95 Set 86061_PM_r_at - -
          Human Genome U95 Set 86061_r_at - -
          Human Genome U133 Set 217548_PM_at - -
          Human Genome U133 Set 217548_at - -
          Human Genome U133 Set 228791_PM_at - -
          Human Genome U133 Set 228791_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 196031 - -
          ABI Human Genome Survey Microarray Version 2.0 234182 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3265143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0