Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03670340_s1
          See other LOC283485 GE Assays ›
          Gene Symbol:
          LOC283485
          Gene Name:
          uncharacterized LOC283485
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03670340_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC283485
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK093862.1 - 1 1951 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF510258.1 - 1 Chr. 13 - 110124753 - 110125234 [-] on Build GRCh38 63
          GenBankmRNA DB348214.1 - 1 Chr. 13 - 110124681 - 110125234 [-] on Build GRCh38 63
          GenBankmRNA AI089021.1 - 1 Chr. 13 - 110124730 - 110125234 [-] on Build GRCh38 63
          GenBankmRNA BF510266.1 - 1 Chr. 13 - 110124770 - 110125234 [-] on Build GRCh38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream long intergenic non-protein coding RNA 396 LINC00396 -68.3kb Sense Chr. 13 - 110053285 - 110054952 on NCBI Build GRCh38
          Downstream collagen type IV alpha 1 chain COL4A1 BSVD,
          RATOR
          23.7kb Anti-sense Chr. 13 - 110148958 - 110307157 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132336 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132355 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16669979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132324 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132347 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132339 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16669968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16669974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132325 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132282 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132320 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132289 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132290 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132288 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16669962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132254 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132338 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16669984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16669969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29581097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09132313 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3692979 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 50766 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3692979 - -
          Agilent Human 1B Microarray G4111A A_32_P713937 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556476_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1556477_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1300008008.hg.1 ENST00000623716.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300008008.hg.1 ENST00000623716.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1556476_at - -
          Human Genome U133 Plus 2.0 Array 1556477_a_at - -
          Human Genome U133 Plus 2.0 Array 1556476_PM_at - -
          Human Genome U133 Plus 2.0 Array 1556477_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556477_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556476_at3 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1828491 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 2350619 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_77588 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1828491 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1828491 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1828491 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0