Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03670597_s1
          See other LOC286382 GE Assays ›
          Gene Symbol:
          LOC286382
          Gene Name:
          uncharacterized LOC286382
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03670597_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC286382
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK094356.1 - 1 1751 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI369795.1 - 1 Chr. 9 - 71059024 - 71059397 [+] on Build GRCh38 80
          GenBankmRNA DB297110.1 - 1 Chr. 9 - 71059042 - 71059581 [+] on Build GRCh38 80
          GenBankmRNA BX095918.1 - 1 190 Chr. 9 - 71059052 - 71059429 [-] on Build GRCh38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 204 MIR204 MIRN204,
          RDICC,
          miRNA204,
          mir-204
          -249.0kb Sense Chr. 9 - 70809975 - 70810084 on NCBI Build GRCh38
          Downstream transmembrane protein 2 TMEM2 622.4kb Sense Chr. 9 - 71683366 - 71769532 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3798739 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P217797 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 1385 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 38488 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3798739 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P217797 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P217797 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 240545_PM_at - -
          Human Genome U133 Plus 2.0 Array 240545_PM_at - -
          Human Genome U133 Plus 2.0 Array 240545_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240545_at11 - -
          Human Genome U95 Set 81638_r_at - -
          Human Genome U95 Set 81638_PM_r_at - -
          Human Genome U95 Set 81636_i_at - -
          Human Genome U95 Set 81636_PM_i_at - -
          Human Genome U133 Set 240545_PM_at - -
          Human Genome U133 Set 240545_at - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1884184 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_79506 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_79506 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1884184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0