Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03670737_m1
          See other LOC284628 GE Assays ›
          Gene Symbol:
          LOC284628
          Gene Name:
          uncharacterized LOC284628
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03670737_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC284628
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK094692.1 - 1 - 2 272 57
          DA148243.1 - 2 - 3 238 57
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA164101.1 - 1 - 2 272 Chr. 1 - 1600439 - 1607180 [+] on Build GRCh38 57

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream fibronectin type III domain containing 10 FNDC10 C1orf233 -343.0 Anti-sense Chr. 1 - 1598008 - 1600096 on NCBI Build GRCh38
          Downstream mindbomb E3 ubiquitin protein ligase 2 MIB2 ZZANK1,
          ZZZ5
          7.5kb Sense Chr. 1 - 1615188 - 1630610 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007133 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007129 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20007117 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 37335 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 536978 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 945877 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 851329 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 657058 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 40253 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 264995 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3751230 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46812 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21686622 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3751230 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P21686622 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P03808092 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21686616 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21686619 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21686614 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21686622 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03808090 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03808097 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03808092 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 83386 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 9121 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 17619 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 14417 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10003197 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21686622 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21686623 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03808090 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03808092 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21686614 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10003209 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03808090 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21686622 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03808092 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21686614 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc