Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03670802_m1
          See other LOC202025 GE Assays ›
          Gene Symbol:
          LOC202025
          Gene Name:
          uncharacterized LOC202025
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03670802_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC202025
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AL713660.1 - 1 - 2 1779 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DR001734.1 - 1 - 2 78 Chr. 4 - 10069222 - 10071318 [-] on Build GRCh38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 2 member 9 SLC2A9 GLUT9,
          GLUTX,
          UAQTL2,
          URATv1
          -27.8kb Sense Chr. 4 - 9771125 - 10040248 on NCBI Build GRCh38
          Downstream WD repeat domain 1 WDR1 AIP1,
          HEL-S-52,
          NORI-1
          1.3kb Sense Chr. 4 - 10074339 - 10116983 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15545237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15545239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15545241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920722 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23073002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23072999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23073008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15545238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23073013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02920716 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 24296 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 92933 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 255337 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 277243 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 86428 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3447441 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P282377 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 28125 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15054 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P16586247 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3447441 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P282377 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P16586247 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P282377 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16586249 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16586252 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P23441417 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P16586249 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 236079_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0400010032.hg.1 ENST00000561486.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400010032.hg.1 ENST00000561486.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400010032.hg.1 ENST00000561486.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400010032.hg.1 ENST00000561486.1 Ensembl
          Human Genome U133 Plus 2.0 Array 236079_PM_at - -
          Human Genome U133 Plus 2.0 Array 236079_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236079_at2 - -
          Human Genome U95 Set 46877_at - -
          Human Genome U95 Set 46877_PM_at - -
          Human Genome U133 Set 236079_at - -
          Human Genome U133 Set 236079_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 228524 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0