Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03670936_m1
          See other LACTB2-AS1 GE Assays ›
          Gene Symbol:
          LACTB2-AS1
          Gene Name:
          LACTB2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 70608577 - 70663279 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03670936_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LACTB2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038881.1 2 - 3 342 69
          GenBank mRNA AL833348.1 - 2 - 3 342 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream lactamase beta 2 LACTB2 CGI-83 Anti-sense Chr. 8 - 70637266 - 70669212 on NCBI Build GRCh38
          Downstream XK related 9 XKR9 XRG9 16.6kb Sense Chr. 8 - 70669365 - 71066635 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800007923.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000465.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201777 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201743 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201742 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201745 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16104680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26484848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06201776 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 104289 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 77102 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 750804 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 813546 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 441286 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 165306 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 193607 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 154990 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 764989 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 279747 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 179920 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 542271 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 938474 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3877992 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14961 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18346358 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P38439424 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P38439424 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3877992 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3877992 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18346358 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P38439424 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 111507 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38439424 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01938972 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16521198 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38439437 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18346393 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P26112018 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16525326 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38439427 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18346388 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01938978 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18346358 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18346358 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P38439424 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01938972 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P16525326 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01938978 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P38439437 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1559264_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR08006533.hg.1 ENST00000499227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006533.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006533.hg.1 NR_038881.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08006533.hg.1 TC08000465.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08006535.hg.1 ENST00000499227.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006535.hg.1 NR_038881.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08006535.hg.1 TC08000465.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518152.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519167.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 ENST00000519358.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007923.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154641.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154641.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154641.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154641.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154641.hg.1 TC0800007923.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800154641.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154641.hg.1 ENST00000518553.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154643.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154643.hg.1 NR_038881.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154643.hg.1 TC0800007923.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800154643.hg.1 ENST00000499227.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154643.hg.1 NR_038881.1 Refseq
          Human Genome U133 Plus 2.0 Array 1559264_at - -
          Human Genome U133 Plus 2.0 Array 1559264_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559264_at5 - -
          Human Genome U95 Set 79904_PM_at - -
          Human Genome U95 Set 79904_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036683 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036683 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0