Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03671063_s1
          See other CBX3P2 GE Assays ›
          Gene Symbol:
          CBX3P2
          Gene Name:
          chromobox 3 pseudogene 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 2652170 - 2655395 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03671063_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CBX3P2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033754.2 2 1163 84
          GenBank mRNA BC018088.1 - 1 751 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB566734.1 - 1 209 Chr. 18 - 2652170 - 2652644 [-] on Build GRCh38 84
          GenBankmRNA BX329736.2 - 1 Chr. 18 - 2652175 - 2653070 [+] on Build GRCh38 84
          GenBankmRNA DB340383.1 - 1 Chr. 18 - 2652171 - 2652707 [+] on Build GRCh38 84
          GenBankmRNA DB565545.1 - 1 192 Chr. 18 - 2652170 - 2652627 [-] on Build GRCh38 84
          GenBankmRNA AA348737.1 - 1 117 Chr. 18 - 2652249 - 2652552 [-] on Build GRCh38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream NDC80,
          kinetochore complex component
          NDC80 HEC,
          HEC1,
          HsHec1,
          KNTC2,
          TID3,
          hsNDC80
          -35.5kb Anti-sense Chr. 18 - 2571511 - 2616635 on NCBI Build GRCh38
          Downstream structural maintenance of chromosomes flexible hinge domain containing 1 SMCHD1 2.7kb Anti-sense Chr. 18 - 2655887 - 2805017 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800007899.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000284.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10509365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17032367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31099847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31099841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31099852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10509363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10509362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17032369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31099854 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 586192 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P94221 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 31412 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 10257 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3606465 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P94221 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P94221 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40931692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1557488_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18003336.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18003336.hg.1 TC18000284.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18003337.hg.1 NR_033754.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18003337.hg.1 TC18000284.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1800154035.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154035.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154035.hg.1 TC1800007899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800154035.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154035.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154039.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154039.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154039.hg.1 TC1800007899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800154039.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154039.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154036.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154036.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154036.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154036.hg.1 TC1800007899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800154036.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154036.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154036.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154040.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154040.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154040.hg.1 TC1800007899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800154040.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154040.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154038.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154038.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154038.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154038.hg.1 TC1800007899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800154038.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154038.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154038.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154037.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154037.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154037.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800154037.hg.1 TC1800007899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800154037.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154037.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800154037.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000579647.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 ENST00000583365.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 NR_033754.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007899.hg.1 NR_033754.2 Refseq
          Human Genome U133 Plus 2.0 Array 1557488_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557488_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557488_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010114 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010114 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1672586 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline