Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03671216_m1
          See other LOC255177 GE Assays ›
          Gene Symbol:
          LOC255177
          Gene Name:
          uncharacterized LOC255177
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03671216_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC255177
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC031230.1 - 2 - 3 1637 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream C2 calcium dependent domain containing 4B C2CD4B FAM148B,
          NLF2
          -31.6kb Sense Chr. 15 - 62163535 - 62165732 on NCBI Build GRCh38
          Downstream golgin A2 pseudogene 11 GOLGA2P11 32.1kb Sense Chr. 15 - 62242529 - 62248795 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16796503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16796501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196464 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16796496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16796502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16796494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703921 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16796491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09703854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30196533 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 550067 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 135502 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 772675 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 570265 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3549874 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P256636 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 24117 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 36865 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3549874 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P256636 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03044433 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P256636 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 31392 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03044433 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03044426 - -
          Agilent Human 1B Microarray G4111A A_32_P476510 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03044433 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1562501_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009674.hg.1 ENST00000559985.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1562501_PM_at - -
          Human Genome U133 Plus 2.0 Array 1562501_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562501_at6 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229106 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc