Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03671220_s1
          See other LOC441455 GE Assays ›
          Gene Symbol:
          LOC441455
          Gene Name:
          makorin ring finger protein 1 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 96725821 - 96727467 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03671220_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC441455
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026792.1 1 1260 88
          GenBank mRNA BC031344.1 - 1 351 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM041694.1 - 1 Chr. 9 - 96726730 - 96727393 [-] on Build GRCh38 88
          GenBankmRNA BM041024.1 - 1 352 Chr. 9 - 96726730 - 96727481 [+] on Build GRCh38 88
          GenBankmRNA BG980941.1 - 1 Chr. 9 - 96726959 - 96727288 [-] on Build GRCh38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream AhpC/TSA antioxidant enzyme domain containing 1 AAED1 C9orf21 -71.4kb Anti-sense Chr. 9 - 96641251 - 96655331 on NCBI Build GRCh38
          Downstream zinc finger protein 510 ZNF510 27.1kb Anti-sense Chr. 9 - 96754549 - 96778549 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008120.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000483.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178326 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178315 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178290 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06836159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27178304 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 36477 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P122402 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P122402 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 13884 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 35327 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P122402 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P122402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836170 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178318 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16244639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836179 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16244639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178318 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27178315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06836170 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P122402 - -
          Agilent Human 1B Microarray G4111A A_32_P122402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836182 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836155 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836177 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836171 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836168 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836174 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836181 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836151 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836160 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16244645 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836166 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836153 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836186 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836164 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836149 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836172 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836162 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06836184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09005153.hg.1 NR_026792.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005153.hg.1 TC09000483.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154434.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154434.hg.1 TC0900008120.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154434.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008120.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154439.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154439.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154439.hg.1 TC0900008120.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154439.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154439.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154435.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154435.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154435.hg.1 TC0900008120.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154435.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154435.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154436.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154436.hg.1 TC0900008120.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154436.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154437.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154437.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154437.hg.1 TC0900008120.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154437.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154437.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154438.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154438.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154438.hg.1 NR_026792.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900154438.hg.1 TC0900008120.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900154438.hg.1 ENST00000445872.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154438.hg.1 ENST00000602289.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900154438.hg.1 NR_026792.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038991 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038992 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038992 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038991 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 156066 - -
          ABI Human Genome Survey Microarray Version 2.0 371288 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3278170 NR_026792.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3278170 BC031344.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0