Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03671649_s1
          See other PP13439 GE Assays ›
          Gene Symbol:
          PP13439
          Gene Name:
          uncharacterized LOC100128046
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03671649_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PP13439
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF318333.1 - 2 2049 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC102724479 LOC102724479 -322.6kb Anti-sense Chr. 3 - 171460495 - 171469673 on NCBI Build GRCh38
          Downstream transmembrane protein 212 TMEM212 49.4kb Anti-sense Chr. 3 - 171843339 - 171859318 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767389 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15511223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15511230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15511228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15511234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767357 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767324 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767379 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15511225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15511224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767320 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767358 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767347 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767384 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15511233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767373 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767391 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02767340 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22907190 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3334843 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P93341 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 2128 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60411 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P93341 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P93341 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001723263_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001726081_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001726374_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1569660_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0300013125.hg.1 ENST00000331659.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013125.hg.1 ENST00000331659.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300013125.hg.1 ENST00000331659.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300013125.hg.1 ENST00000331659.2 Ensembl
          Human Genome U133 Plus 2.0 Array 1569660_PM_at - -
          Human Genome U133 Plus 2.0 Array 1569660_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569660_at8 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0