Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
236166_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
242852_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011668.hg.1 |
ENST00000569008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011668.hg.1 |
ENST00000569008.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
242852_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
236166_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
236166_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
242852_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236166_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
242852_at9 |
- |
- |
Human Genome U95 Set |
52515_PM_at |
- |
- |
Human Genome U95 Set |
52515_at |
- |
- |
Human Genome U95 Set |
59269_PM_at |
- |
- |
Human Genome U95 Set |
59269_at |
- |
- |
Human Genome U133 Set |
242852_at |
- |
- |
Human Genome U133 Set |
236166_PM_at |
- |
- |
Human Genome U133 Set |
236166_at |
- |
- |
Human Genome U133 Set |
242852_PM_at |
- |
- |