Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03672789_s1
          See other LOC100129785 GE Assays ›
          Gene Symbol:
          LOC100129785
          Gene Name:
          uncharacterized LOC100129785
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03672789_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129785
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK125285.1 - 1 3045 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream G protein-coupled receptor 107 GPR107 GCDRP,
          LUSTR1,
          bA138E2.2
          Sense Chr. 9 - 130053706 - 130140169 on NCBI Build GRCh38
          Downstream uncharacterized LOC401554 LOC401554 18.4kb Anti-sense Chr. 9 - 130140610 - 130144219 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27326998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27327004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27327019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27327010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06979574 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 26212 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 4896 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 315030 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 375322 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 965792 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 690181 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3392004 - -
          Agilent Human Genome CGH Microarray 44B A_14_P127479 - -
          Agilent Human Genome CGH Microarray 44A A_14_P127479 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53111 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3392004 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P127479 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P127479 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P127479 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 143079 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 16169 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P127479 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02185120 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02185115 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P127479 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8158626 - -
          Affymetrix Human Gene 1.1 ST Array 8158625 - -
          Affymetrix Human Transcriptome Array 2.0 TC0900008947.hg.1 ENST00000411256.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008947.hg.1 ENST00000411256.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3260546 AK125285.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3260546 AK125285.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0