Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03672866_s1
          See other LOC652990 GE Assays ›
          Gene Symbol:
          LOC652990
          Gene Name:
          uncharacterized LOC652990
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03672866_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC652990
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK126413.1 - 1 248 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE061880.1 - 1 190 Chr. 6 - 44095173 - 44095520 [-] on Build GRCh38 92
          GenBankmRNA BE062059.1 - 1 190 Chr. 6 - 44095173 - 44095519 [-] on Build GRCh38 92
          GenBankmRNA DB263562.1 - 1 248 Chr. 6 - 44095013 - 44095585 [-] on Build GRCh38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC101929705 LOC101929705 -18.3kb Sense Chr. 6 - 43995723 - 44074652 on NCBI Build GRCh38
          Downstream mitochondrial ribosomal protein L14 MRPL14 L14mt,
          L32mt,
          MRP-L14,
          MRP-L32,
          MRPL32,
          RMPL32,
          RPML32
          18.1kb Sense Chr. 6 - 44113454 - 44127491 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15811654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15811651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24895063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15811652 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24895015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24895033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15811656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24895039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24895068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15811662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15811658 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699887 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699895 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24895048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699907 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04699902 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118571 AK097146.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118571 EF186024.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118571 NM_152424.3 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 204401 BC039896.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 204401 BC045825.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 204401 CR596589.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 204401 CR749471.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 204401 NM_005156.4 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 835274 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3327046 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P15626554 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3327046 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17528118 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15626554 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15626554 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000419707.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000419707.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000419707.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000502812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000502812.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000511517.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000511517.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000511517.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000512508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000512508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000512508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000514784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009761.hg.1 ENST00000514784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000419707.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000419707.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000419707.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000502812.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000502812.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000509080.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000511517.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000511517.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000511517.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000512508.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000512508.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000512508.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000514784.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500009761.hg.1 ENST00000514784.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix