Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03672967_s1
          See other ERV18-1 GE Assays ›
          Gene Symbol:
          ERV18-1 UNC13C
          Gene Name:
          endogenous retrovirus group 18 member 1
          unc-13 homolog C
          Gene Aliases:
          -
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.15: 53837573 - 54628608 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03672967_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERV18-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK126787.1 - 1 1952 99
          Gene Symbol: UNC13C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017022223.1 XP_016877712.1 1 1090 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream WD repeat domain 72 WDR72 AI2A3 -147.9kb Anti-sense Chr. 15 - 53513741 - 53762878 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_017022223.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          chemical synaptic transmission
          intracellular signal transduction
          diacylglycerol binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157952 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665399 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665462 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16792016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16792009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157992 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665456 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665448 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30157988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30158004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09665408 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 507454 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3274179 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14533 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3274179 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500007293.hg.1 ENST00000624468.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007293.hg.1 ENST00000624468.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0