Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03673601_s1
          See other LOC100129596 GE Assays ›
          Gene Symbol:
          LOC100129596
          Gene Name:
          uncharacterized LOC100129596
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03673601_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129596
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK130913.1 - 1 345 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 517 ZNF517 -9.9kb Anti-sense Chr. 8 - 144798876 - 144813033 on NCBI Build GRCh38
          Downstream zinc finger protein 7 ZNF7 HF.16,
          KOX4,
          zf30
          6.8kb Anti-sense Chr. 8 - 144827500 - 144848070 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537347 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537254 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537263 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26824548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16150673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06537260 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 152471 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 7336 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 11841 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 53769 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 19148 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 144457 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 66023 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 25310 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 11824 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 17788 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 39736 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 48155 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 275059 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 352848 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 153744 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 509293 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 737873 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 534484 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 592916 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 195919 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 924305 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 731005 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 169324 NM_000833.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 169324 U09002.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 287824 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 11704 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 837743 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 376147 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 931279 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 750965 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 21127 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 357469 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 675876 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 961419 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 581137 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 202557 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 352121 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AB032427.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AB073669.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AB100308.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AF258465.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AF263523.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AF279673.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AJ296305.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 AL834192.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 BC117426.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 DQ059644.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 DQ059645.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 DQ059646.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 NM_021625.3 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 145823 NM_147204.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 95581 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 AK026277.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 AK291496.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 BC007119.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 BC025367.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 BC044213.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 BC056876.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 BX648605.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 CR617422.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 CR625945.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 183606 NM_024808.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 278988 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 97106 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28336 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18524365 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18524339 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P59969097 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02044388 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P26374032 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18524473 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02044388 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18524339 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18524473 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3364964 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18524365 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18524407 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P02044388 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18524439 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524443 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524339 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02044373 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524580 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524473 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524365 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02044405 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524439 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02044309 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524426 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18524407 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02044339 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02044388 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 168191 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 86956 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 120033 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 113033 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 130603 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 96005 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 18312 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 67803 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 86381 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044360 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044386 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38620770 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044353 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044346 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044283 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16712599 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044287 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524354 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044388 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524582 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044373 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524585 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044301 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044348 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524473 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P26372344 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524579 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P26373027 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044309 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524580 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524351 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524365 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02044405 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18524339 - -
          Agilent Human 1B Microarray G4111A A_32_P383873 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02044388 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18524365 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18524351 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02044373 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02044309 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18524339 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P16712599 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P38620770 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18524473 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3252960 AK130913.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0