Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03673860_s1
          See other LOC440934 GE Assays ›
          Gene Symbol:
          LOC440934
          Gene Name:
          uncharacterized LOC440934
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 222318471 - 222357910 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03673860_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC440934
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_136638.1 1 356 90
          NR_136642.1 1 356 90
          GenBank mRNA BC008048.2 - 1 355 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI711783.1 - 1 133 Chr. 2 - 222318701 - 222342541 [+] on Build GRCh38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream coiled-coil domain containing 140 CCDC140 -13.3kb Sense Chr. 2 - 222298147 - 222305217 on NCBI Build GRCh38
          Downstream sphingosine-1-phosphate phosphatase 2 SGPP2 SPP2,
          SPPase2
          104.7kb Sense Chr. 2 - 222424052 - 222562622 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15386592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01917201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22042896 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 415093 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 39285 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 6073 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3358005 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8048713 - -
          Affymetrix HT HG-U133+ PM Array Plate 230844_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 244159_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000590427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000590427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010895.hg.1 ENST00000590427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000424010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000429475.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000439237.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000446532.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000555502.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000587192.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000589724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000590427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000590427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010895.hg.1 ENST00000590427.1 Ensembl
          Human Genome U133 Plus 2.0 Array 230844_PM_at - -
          Human Genome U133 Plus 2.0 Array 244159_at - -
          Human Genome U133 Plus 2.0 Array 230844_at - -
          Human Genome U133 Plus 2.0 Array 244159_PM_at - -
          Human Genome U133 Plus 2.0 Array 238727_at - -
          Human Genome U133 Plus 2.0 Array 238727_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244159_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230844_at3 - -
          Human Genome U95 Set 44818_PM_s_at - -
          Human Genome U95 Set 49243_PM_at - -
          Human Genome U95 Set 44818_s_at - -
          Human Genome U95 Set 49243_at - -
          Human Genome U95 Set 57364_PM_at - -
          Human Genome U95 Set 57364_at - -
          Human Genome U95 Set 54482_PM_at - -
          Human Genome U95 Set 54482_at - -
          Human Genome U95 Set 80151_at - -
          Human Genome U95 Set 80151_PM_at - -
          Human Genome U133 Set 238727_PM_at - -
          Human Genome U133 Set 230844_PM_at - -
          Human Genome U133 Set 230844_at - -
          Human Genome U133 Set 244159_PM_at - -
          Human Genome U133 Set 244159_at - -
          Human Genome U133 Set 238727_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1741507 BC008048.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1741507 BC008048.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1741507 BC008048.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1741507 BC008048.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0