Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03675203_s1
          See other LINC01165 GE Assays ›
          Gene Symbol:
          LINC01165 LOC107984282
          Gene Name:
          long intergenic non-protein coding RNA 1165
          uncharacterized LOC107984282
          Gene Aliases:
          -
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.10: 132508444 - 132536923 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03675203_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01165
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK097584.1 - 1 1046 61
          Gene Symbol: LOC107984282
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017017028.1 XP_016872517.1 2 3331 61

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chromosome 10 open reading frame 91 C10orf91 bA432J24.4 -72.5kb Anti-sense Chr. 10 - 132445210 - 132448321 on NCBI Build GRCh38
          Downstream inositol polyphosphate-5-phosphatase A INPP5A 5PTASE 15.4kb Anti-sense Chr. 10 - 132537779 - 132783480 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27980457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27980420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27980414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16401294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16401305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16401312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27980459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16401313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27980454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589263 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589284 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27980453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16401301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589266 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07589295 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 302448 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 257181 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 239207 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 327989 AK054626.1 genbank_mrna_human
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375259 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 41757 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3375259 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02376928 - -
          Agilent Human 1B Microarray G4111A A_32_P496315 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001719712_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001715157_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001716061_at - -
          Affymetrix Human Gene 1.0 ST Array 7937105 - -
          Affymetrix Human Gene 1.1 ST Array 7937104 - -
          Affymetrix HT HG-U133+ PM Array Plate 1564490_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1000012352.hg.1 ENST00000445190.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012352.hg.1 ENST00000445190.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1564490_at - -
          Human Genome U133 Plus 2.0 Array 1564490_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564490_at1 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1870717 AK097584.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1870717 AK097584.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1870717 AK097584.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 7400259 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_110243 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1870717 AK097584.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1870717 AK097584.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1870717 AK097584.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0