Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03675739_s1
          See other HCRP1 GE Assays ›
          Gene Symbol:
          HCRP1
          Gene Name:
          hepatocellular carcinoma-related HCRP1
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03675739_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HCRP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK025343.1 - 1 151 102
          AY465895.1 - 1 150 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX471748.1 - 1 150 Chr. 6 - 42358793 - 42359443 [+] on Build GRCh38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transcriptional regulating factor 1 TRERF1 BCAR2,
          HSA277276,
          RAPA,
          TREP132,
          TReP-132,
          dJ139D8.5
          Anti-sense Chr. 6 - 42224933 - 42452216 on NCBI Build GRCh38
          Downstream ubiquitin protein ligase E3 component n-recognin 2 UBR2 C6orf133,
          bA49A4.1,
          dJ242G1.1,
          dJ392M17.3
          203.2kb Sense Chr. 6 - 42563905 - 42693505 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the AY465895.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3405491 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16236 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3405491 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001725456_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001722898_at - -
          Affymetrix Human Gene 1.0 ST Array 8119526 - -
          Affymetrix Human Gene 1.1 ST Array 8119525 - -
          Affymetrix HT HG-U133+ PM Array Plate 216174_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 216176_PM_at - -
          Affymetrix Human Genome U219 Array 11760722_at - -
          Affymetrix Human Genome U219 Array 11760722_PM_at - -
          Human Genome U133 Plus 2.0 Array 216176_at - -
          Human Genome U133 Plus 2.0 Array 216174_PM_at - -
          Human Genome U133 Plus 2.0 Array 216174_at - -
          Human Genome U133 Plus 2.0 Array 216176_PM_at - -
          Human Genome U133A 2.0 Array 216176_PM_at - -
          Human Genome U133A 2.0 Array 216174_at - -
          Human Genome U133A 2.0 Array 216174_PM_at - -
          Human Genome U133A 2.0 Array 216176_at - -
          AffymetrixPathwork Diagnostics Pathchip pw567299 - -
          AffymetrixPathwork Diagnostics Pathchip pw561618 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216176_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216174_at9 - -
          Human Genome U133 Set 216176_PM_at - -
          Human Genome U133 Set 216176_at - -
          Human Genome U133 Set 216174_PM_at - -
          Human Genome U133 Set 216174_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3252608 AK025343.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3252608 AY465895.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0