Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03677234_m1
          See other DKFZP434L187 GE Assays ›
          Gene Symbol:
          DKFZP434L187
          Gene Name:
          uncharacterized LOC26082
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 30196036 - 30214540 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03677234_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DKFZP434L187
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026771.1 2 - 3 128 95
          GenBank mRNA BC130408.1 - 2 - 3 128 95
          BC130410.1 - 2 - 3 128 95
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX283347.1 - 2 - 3 89 Chr. 15 - 30196076 - 30210794 [+] on Build GRCh38 95
          GenBankmRNA BI114068.1 - 2 - 3 89 Chr. 15 - 30196076 - 30210915 [+] on Build GRCh38 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105370746 LOC105370746 -19.3kb Anti-sense Chr. 15 - 30169985 - 30179938 on NCBI Build GRCh38
          Downstream CHRNA7 (exons 5-10) and FAM7A (exons A-E) fusion CHRFAM7A CHRNA7,
          CHRNA7-DR1,
          D-10
          143.2kb Anti-sense Chr. 15 - 30357770 - 30393849 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500006707.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000164.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09567418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30046423 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 243216 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 973464 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 129085 AF141315.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 129085 AK225668.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 129085 BC119639.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 129085 BC119640.2 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 129085 NM_016161.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 405497 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 228182 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 756026 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 187843 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 501447 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 571775 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 810611 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 890168 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 242443 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 220678 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 33755 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 310056 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 232249 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 151234 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 313189 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 392283 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 20221 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133921 AK090413.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133921 BC033745.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133921 BC040030.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133921 BC101371.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133921 BC101372.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133921 BC141843.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 133921 NM_178026.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 297872 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3328782 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P478512 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 14757 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 56965 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 7946 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P12162649 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P478512 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3328782 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3328782 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P478512 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30046423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30046423 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P478512 - -
          Agilent Human 1B Microarray G4111A A_32_P478512 - -
          Agilent Human 1B Microarray G4111A A_32_P777089 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09562418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7982253 - -
          Affymetrix Human Gene 1.1 ST Array 7982252 - -
          Affymetrix HT HG-U133+ PM Array Plate 230861_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 216596_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1569476_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 ENST00000455163.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15001062.hg.1 ENST00000455163.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15001062.hg.1 NR_026771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15001062.hg.1 TC15000164.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15001063.hg.1 NR_026771.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15001063.hg.1 TC15000164.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500146523.hg.1 ENST00000455163.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146523.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146523.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500146523.hg.1 TC1500006707.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500146523.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500146523.hg.1 ENST00000455163.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146523.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146525.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146525.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500146525.hg.1 TC1500006707.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500146525.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146525.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 ENST00000455163.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006707.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500146524.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146524.hg.1 NR_026771.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500146524.hg.1 TC1500006707.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500146524.hg.1 ENST00000562624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500146524.hg.1 NR_026771.1 Refseq
          Human Genome U133 Plus 2.0 Array 216596_PM_at - -
          Human Genome U133 Plus 2.0 Array 230861_PM_at - -
          Human Genome U133 Plus 2.0 Array 1569476_PM_at - -
          Human Genome U133 Plus 2.0 Array 1569476_at - -
          Human Genome U133 Plus 2.0 Array 216596_at - -
          Human Genome U133 Plus 2.0 Array 230861_at - -
          Human Genome U133A 2.0 Array 216596_PM_at - -
          Human Genome U133A 2.0 Array 216596_at - -
          AffymetrixPathwork Diagnostics Pathchip pw514890 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569476_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230861_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216596_at4 - -
          Human Genome U95 Set 62721_at - -
          Human Genome U95 Set 62721_PM_at - -
          Human Genome U133 Set 216596_PM_at - -
          Human Genome U133 Set 216596_at - -
          Human Genome U133 Set 230861_PM_at - -
          Human Genome U133 Set 230861_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052702 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052701 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052701 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052702 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 174074 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1669413 NR_026771.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1669413 AL117445.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1669413 NR_026771.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1669413 AL117445.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1669413 NR_026771.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1669413 AL117445.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1669413 NR_026771.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1669413 AL117445.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1669413 NR_026771.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1669413 AL117445.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0