Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03677616_mH
          See other ANKRD65 GE Assays ›
          Gene Symbol:
          ANKRD65
          Gene Name:
          ankyrin repeat domain 65
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1418420 - 1422561 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03677616_mH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANKRD65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243535.1 NP_001230464.1 2 - 3 522 103
          XM_017001325.1 XP_016856814.1 1 - 2 1512 103
          GenBank mRNA BC018095.1 - 2 - 3 342 103
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM465285.1 - 300 Chr. - - [-] on Build 103
          GenBankmRNA CD387703.1 - 3 - 4 375 Chr. 1 - 1419068 - 2221025 [-] on Build GRCh38 103
          GenBankmRNA BE263901.1 - 1 - 2 441 Chr. 1 - 1419416 - 1421236 [-] on Build GRCh38 103
          GenBankmRNA HY098541.1 - 2 - 3 402 Chr. 1 - 1419482 - 1421324 [-] on Build GRCh38 103
          GenBankmRNA BI116776.1 - 1 - 2 141 Chr. 1 - 1418905 - 1420936 [-] on Build GRCh38 103
          GenBankmRNA BM142089.1 - 2 - 3 308 Chr. 1 - 1419313 - 1421230 [-] on Build GRCh38 103
          GenBankmRNA BF341239.1 - 2 - 3 304 Chr. 1 - 1419138 - 1421226 [-] on Build GRCh38 103
          GenBankmRNA BI755193.1 - 1 - 2 465 Chr. 1 - 1419153 - 1421260 [-] on Build GRCh38 103
          GenBankmRNA BQ948384.1 - 143 Chr. - - [-] on Build 103
          GenBankmRNA BE795877.1 - 1 - 2 427 Chr. 1 - 1419188 - 1421223 [-] on Build GRCh38 103
          GenBankmRNA BE798505.1 - 2 - 3 364 Chr. 1 - 1419135 - 1421260 [-] on Build GRCh38 103
          GenBankmRNA CK819891.1 - 2 - 3 324 Chr. 1 - 1419300 - 1421246 [-] on Build GRCh38 103
          GenBankmRNA BI770529.1 - 2 - 3 336 Chr. 1 - 1419212 - 1421257 [-] on Build GRCh38 103
          GenBankmRNA BM451685.1 - 81 Chr. - - [-] on Build 103
          GenBankmRNA DB493759.1 - 2 - 3 342 Chr. 1 - 1419386 - 1421264 [-] on Build GRCh38 103
          GenBankmRNA BM353987.1 - 2 - 3 308 Chr. 1 - 1419310 - 1421230 [-] on Build GRCh38 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mitochondrial ribosomal protein L20 MRPL20 L20mt,
          MRP-L20
          -10.6kb Sense Chr. 1 - 1401896 - 1407313 on NCBI Build GRCh38
          Downstream transmembrane protein 88B TMEM88B 5.1kb Anti-sense Chr. 1 - 1426128 - 1427787 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline